missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-B7 Monoclonal Antibody (BB7.1), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
158.00€ - 398.00€
Spécification
Antigène | HLA-B7 |
---|---|
Clone | BB7.1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16370784
|
Affymetrix eBioscience
25-5917-42 |
100 Tests |
398.00€
100 pièces |
Expédition estimée: 10-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16360784
|
Affymetrix eBioscience
25-5917-41 |
25 Tests |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
Mouse Monoclonal Antibody
HLA-B7 is one of the polymorphic proteins of human MHC Class I found on all nucleated cells. This surface protein is a heterodimer consisting of the specific transmembrane HLA-B chain paired with Beta-2 microglobulin. Classical MHC Class I molecules play a central role in adaptive immunity by presenting to T cells antigenic peptides derived either from the cell itself (classical presentation) or extracellular antigens (cross-presentation). The ligands of MHC class I include T cell receptor (TCR) and a costimulatory molecule CD8. Hundreds of HLA-B alleles have been described. Typing for these polymorphisms is routinely done for transplantation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Spécification
HLA-B7 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P01889 | |
HLA-B | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
BB7.1 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
3106 | |
Papain solubilized HLA-B7 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
HLA-B |