missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25142-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
HNRPA3 contains 2 RRM (RNA recognition motif) domains and plays a role in cytoplasmic trafficking of RNA. It binds to the cis-acting response element, A2RE and may be involved in pre-mRNA splicing.Spécification
HNRNPA3 | |
Polyclonal | |
Unconjugated | |
HNRNPA3 | |
2610510D13Rik, D10S102, FBRNP, hnRNP A3, HNRNPA3, HNRPA3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
220988, 229279 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.63 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51991, Q8BG05 | |
HNRNPA3 | |
HNRNPA3 Fusion Protein Ag18248 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |