Learn More
HOOK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15457-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16834644
  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
Description
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
Spécification
Spécification
HOOK3
Polyclonal
Unconjugated
HOOK3
h hook3, hHK3, HK3, hook homolog 3 (Drosophila), HOOK3, Protein Hook homolog 3
Rabbit
Antigen Affinity Chromatography
RUO
306548, 320191, 84376
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86VS8, Q8BUK6
Hook3
HOOK3 Fusion Protein Ag7724
20 μL
Primary
Human, Mouse, Rat