Learn More
HTR5A Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28221-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17271396
  • 140.00€ / 20µL
Expédition estimée: 17-06-2024
Description
The HTR5A antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human HTR5A. This antibody recognizes human, mouse antigen. The HTR5A antibody has been validated for the following applications: IHC,ELISA analysis.
Spécification
Spécification
HTR5A
Polyclonal
Unconjugated
20 μL
3361
Liquid
Immunohistochemistry, ELISA
550 μg/mL
Rabbit
Primary
Human, Mouse