missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13830-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
5-HT7 Receptor is a Serotonin Receptor with several functions, including regulation of the circadian rhythm and mediation of smooth-muscle relaxation in the vasculature and GI tract. This receptor has been implicated in depression, autism, and other neuropsychiatric disorders. Three protein isoforms have been documented: isoform a: 445aa; isoform b: 432aa; isoform d: 479aa. 5-HT7 Receptor has been reported in aorta, brain, heart, placenta, small intestine, spleen, artery, and vessel. An EST has been isolated from a testis library.Spécification
HTR7 | |
Polyclonal | |
Unconjugated | |
HTR7 | |
5 HT 7, 5 HT X, 5 HT7, 5 hydroxytryptamine receptor 7, HTR7, Serotonin receptor 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15566, 3363, 65032 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32304, P32305, P34969 | |
HTR7 | |
HTR7 Fusion Protein Ag4742 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |