Learn More
ID1 Mouse, anti-Human, Unconjugated, Clone: 1H3G11, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67827-1-IG-20UL
Informations supplémentaires : Poids : 0.00020kg
 Afficher plus de versions de ce produit
Code produit. 17271476
  • 140.00€ / 20µL
Expédition estimée: 30-04-2024
pour voir le stock disponible 
Description
Description
ID (inhibitor of DNA binding) proteins contain a helix-loop-helix (HLH) motif and regulate tissue-specific transcription within several cell lineages. They do not bind DNA directly, but inhibit lineage commitment by binding basic helix-loop-helix (bHLH) transcription factors through their HLH motif. ID1 proteins lack a basic DNA-binding domain but are able to form heterodimers with other HLH proteins, thereby inhibiting DNA binding. ID proteins contribute to cell growth, senescence, differentiation, and angiogenesis. Inhibitor of differentiation or DNA binding-1 (ID1) interacts with the transactivation domain of p65 (p65TAD) both in vitro and in vivo. The molecular weight of ID1 and p65 complex is about 45-50 kDa (PMID: 22592405 ).
Spécification
Spécification
ID1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
Western Blot, ELISA
1H3G11
Unconjugated
20 μL
3397