missing translation for 'onlineSavingsMsg'
Learn More

Insulin Mouse anti-Human, Mouse, Clone: 4B6A7, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66198-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837855

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
Spécification

Spécification

Insulin
Monoclonal
1.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P01308
INS
Insulin Fusion Protein Ag8630
150 μL
Primary
Mouse, Human
Antibody
IgG2a
Immunohistochemistry (Paraffin)
4B6A7
Unconjugated
INS
ILPR, INS, Insulin, IRDN
Mouse
Protein A
RUO
3630
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions