missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF3C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14333-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
KIF3C is a microtubule-based anterograde translocator for membranous organelles.Spécification
KIF3C | |
Polyclonal | |
Unconjugated | |
KIF3C | |
KIF3C, kinesin family member 3C, Kinesin like protein KIF3C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16570, 3797, 85248 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14782, O35066, O55165 | |
Kif3c | |
KIF3C Fusion Protein Ag5619 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |