missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14849-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
KLHL14 is a member of the KLHL family.Spécification
KLHL14 | |
Polyclonal | |
Unconjugated | |
KLHL14 | |
kelch like 14 (Drosophila), Kelch like protein 14, KIAA1384, KLHL14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
225266, 364823, 57565 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q69ZK5, Q9P2G3 | |
Klhl14 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |