missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23498-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.Spécification
LAMB1 | |
Polyclonal | |
Unconjugated | |
LAMB1 | |
CLM, LAMB1, Laminin 1 subunit beta, Laminin 10 subunit beta, Laminin 12 subunit beta, Laminin 2 subunit beta, Laminin 6 subunit beta, Laminin 8 subunit beta, Laminin B1 chain, Laminin subunit beta 1, laminin, beta 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16777, 298941, 3912 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02469, P07942 | |
Lamb1 | |
LAMB1 Fusion Protein Ag19182 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |