Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-ERK2 [pThr185/pTyr187] Antibody
A part of the MAPK Pathway, ERK2 is phosphorylated at Thr185 and Tyr187 by the mitogen-activated protein kinase kinases 2 (MAP2K2).
Marque: Thermo Scientific™ PV5210
Code nomenclature Nacres: NA.47
Informations supplémentaires : Poids : 0.00900kg
Description
- A part of the MAPK Pathway, ERK2 is phosphorylated at Thr185 and Tyr187 by the mitogen-activated protein kinase kinases 2 (MAP2K2)
- For use in combination with the LanthaScreen™ ERK2 A375 cell line (K1798) to evaluate inhibitors in medium to high throughput screening applications
Spécification
Tb (Terbium)/Lanthanide Chelates | |
Terbium-labeled pERK2 (pThr185/pTyr187) antibody in HEPES buffered saline. Store at -20°C. | |
Human | |
End Point | |
Ser/Thr Kinases, Tyrosine Kinases |
1 mg | |
340/490, 520 | |
Dry Ice | |
Hu | |
LanthaScreen™ |
For Research Use Only. Not for use in diagnostic procedures.