Learn More
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit
Marque: Invitrogen PV4844
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.50000kg
Description
A part of the JAK/STAT Pathway, STAT1 (Signal Transducer and Activator of Transcription 1) is phosphorylated at Tyr701 by the Janus family of tyrosine kinases.For use in combination with the LanthaScreen™ STAT1 U206 cell line (K1469) and the GFP-STAT1 physiological protein substrate (PV5211) to evaluate inhibitors in medium to high throughput screening applications.
Spécification
Tyrosine kinases | |
Tb (Terbium)/GFP (EmGFP) | |
25 μg | |
Protein Assays and Analysis | |
1 Kit(s) | |
150000 Da | |
384-well Plate | |
Human | |
End Point | |
STAT1, JAK1, JAK2 JH1 JH2, JAK2, TYK2, JAK3, JAK2 JH1 JH2 V617F | |
TR-FRET |
Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay | |
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit, 25μg, Tyrosine Kinases Antigen, Human Species Reactivity, Used For Biochemical Kinase Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays, | |
RUO, RUO | |
Primary | |
Terbium-labeled anti-STAT1 [pTyr701] antibody in HEPES buffered saline. Store at -20°C. | |
Biochemical terbium phospho-specific activity, Cell-based terbium phospho-specific activity (stable) | |
340⁄490, 520 | |
Dry Ice | |
Hu | |
Tyrosine Kinases | |
JAK1, JAK2, JAK3, TYK2 |
For Research Use Only. Not for use in diagnostic procedures.