Learn More
LAYN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 20535-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16830085
  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
Description
Layilin is a receptor for hyaluronate.
Spécification
Spécification
LAYN
Polyclonal
Unconjugated
LAYN
layilin, LAYN
Rabbit
Antigen Affinity Chromatography
RUO
143903, 244864
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UX15, Q8C351
LAYN
LAYN Fusion Protein Ag14508
20 μL
Primary
Mouse, Human