missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LITAF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16797-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Lipopolysaccharide is a potent stimulator of monocytes and macrophages, causing secretion of tumor necrosis factor-alpha (TNF-alpha) and other inflammatory mediators. This gene encodes lipopolysaccharide-induced TNF-alpha factor, which is a DNA-binding protein and can mediate the TNF-alpha expression by direct binding to the promoter region of the TNF-alpha gene.Spécification
LITAF | |
Polyclonal | |
Unconjugated | |
LITAF | |
LITAF, LPS induced TNF alpha factor, p53 induced gene 7 protein, PIG7, SIMPLE, TP53I7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56722, 65161, 9516 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0C0T0, Q99732, Q9JLJ0 | |
Litaf | |
LITAF Fusion Protein Ag10269 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |