missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPPR2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25732-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
LPPR2 is a multi-pass membrane protein and belongs to the PA-phosphatase related phosphoesterase family. LPPR2 shows dynamic expression regulation during brain development and neuronal excitation.Spécification
LPPR2 | |
Polyclonal | |
Unconjugated | |
PLPPR2 | |
LPPR2, PRG 4, PRG4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235044, 64748 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8VCY8, Q96GM1 | |
PLPPR2 | |
LPPR2 Fusion Protein Ag22839 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |