missing translation for 'onlineSavingsMsg'
Learn More

LUC7L2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24202-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832725

  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a protein that contains a C2H2-type zinc finger, coiled-coil region and arginine, serine-rich (RS) domain. A similar protein in mouse interacts with sodium channel modifier 1, and the encoded protein may be involved in the recognition of non-consensus splice donor sites in association with the U1 snRNP spliceosomal subunit. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Spécification

Spécification

LUC7L2
Polyclonal
Unconjugated
LUC7L2
CGI 59, CGI 74, LUC7 like 2 (S. cerevisiae), LUC7B2, LUC7L2
Rabbit
Antigen Affinity Chromatography
RUO
51631
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y383
LUC7L2
LUC7L2 Fusion Protein Ag21135
150 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions