Learn More
LXN Mouse anti-Human, Unconjugated, Clone: 3E5A5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66957-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The LXN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LXN. This antibody recognizes human, pig, rabbit antigen. The LXN antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
LXN | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Pig, Rabbit |
Western Blot, Immunohistochemistry, ELISA | |
3E5A5 | |
Unconjugated | |
20 μL | |
56925 | |
IgG1 |