Learn More
Ly-6G Rat anti-Mouse, BB700, Clone: 1A8, BD Horizon™
Rat Monoclonal Antibody
Marque: BD Biosciences 566453
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00810kg
Description
Ly-6G Monoclonal antibody specifically detects Ly-6G in Mouse samples. It is validated for Flow Cytometry.Spécification
Ly-6G | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
1A8 | |
Brilliant Blue 700 (BB700) | |
25 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |