missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPLAL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16146-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
LYPLAL1 does not exhibit phospholipase nor triacylglycerol lipase activity. It is able to hydrolyze only short chain substrates due to its shallow active site.Spécification
LYPLAL1 | |
Polyclonal | |
Unconjugated | |
LYPLAL1 | |
KIAA1238, LYPLAL1, lysophospholipase like 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
127018, 226791 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UFF7, Q5VWZ2 | |
Lyplal1 | |
LYPLAL1 Fusion Protein Ag9271 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |