missing translation for 'onlineSavingsMsg'
Learn More

MAPK4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26102-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16894625

  • 349.00€ / 150µL
Expédition estimée: 12-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets.
Spécification

Spécification

MAPK4
Polyclonal
Unconjugated
MAPK4
ERK 4, ERK3, Erk4, MAP kinase 4, MAP kinase isoform p63, MAPK 4, MAPK4, p63 MAPK, p63MAPK, PRKM4
Rabbit
Antigen Affinity Chromatography
RUO
225724, 5596
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P31152, Q6P5G0
MAPK4
MAPK4 Fusion Protein Ag22899
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions