missing translation for 'onlineSavingsMsg'
Learn More

Marcksl1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10003-0-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836163

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Controls cell movement by regulating actin cytoskeleton homeostasis and filopodium and lamellipodium formation. When unphosphorylated, induces cell migration. When phosphorylated by MAPK8, induces actin bundles formation and stabilization, thereby reducing actin plasticity, hence restricting cell movement, including neuronal migration. May also affect cancer cell migration. May be involved in coupling the protein kinase C and calmodulin signal transduction systems.
Spécification

Spécification

Marcksl1
Polyclonal
Unconjugated
MARCKSL1
AL022768, AW215397, AW536807, D4Bc1, F52, MacMARCKS, Macs 2, Macs 3, Macs2, Macs3, MARCKS like 1, Marcksl1, Mlp, Mrp
Rabbit
150 μL
Primary
Mouse, Human
Antibody
IgG
ELISA
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P28667, P49006
Marcksl1
Protein A
RUO
17357, 65108
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions