Learn More
MASTL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15739-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The MASTL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MASTL. This antibody recognizes human, mouse antigen. The MASTL antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
MASTL | |
Polyclonal | |
Unconjugated | |
150 μL | |
84930 | |
IgG |
Immunohistochemistry, ELISA | |
247 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |