Learn More
MASTL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15739-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16825054
  • 399.00€ / 150µL
Expédition estimée: 29-05-2024
Description
The MASTL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MASTL. This antibody recognizes human, mouse antigen. The MASTL antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
Spécification
MASTL
Polyclonal
Unconjugated
150 μL
84930
IgG
Immunohistochemistry, ELISA
247 μg/mL
Rabbit
Primary
Human, Mouse