Learn More
MASTL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15739-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16835054
  • 140.00€ / 20µL
Expédition estimée: 28-05-2024
pour voir le stock disponible 
Description
Description
The MASTL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MASTL. This antibody recognizes human, mouse antigen. The MASTL antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
Spécification
MASTL
Polyclonal
Unconjugated
20 μL
84930
IgG
Immunohistochemistry, ELISA
247 μg/mL
Rabbit
Primary
Human, Mouse