Learn More
MBD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15961-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835374
  • 140.00€ / 20µL
Expédition estimée: 15-05-2024
Description
MBD5 binds to heterochromatin but does not interact with either methylated or unmethylated DNA (in vitro).
Spécification
Spécification
MBD5
Polyclonal
Unconjugated
MBD5
KIAA1461, MBD5, MRD1
Rabbit
Antigen Affinity Chromatography
RUO
109241, 311026, 55777
-20°C
Liquid
Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B1AYB6, Q9P267
MBD5
MBD5 Fusion Protein Ag8728
20 μL
Primary
Human, Rat, Mouse