missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24207-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.Spécification
MBL2 | |
Polyclonal | |
Unconjugated | |
MBL2 | |
COLEC1, Collectin 1, HSMBPC, Man binding protein, Mannose binding lectin, Mannose binding protein C, MBL, MBL2, MBP, MBP C, MBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4153 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11226 | |
MBL2 | |
MBL2 Fusion Protein Ag21508 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |