missing translation for 'onlineSavingsMsg'
Learn More

MBL2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24207-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16892725

  • 399.00€ / 150µL
Expédition estimée: 03-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
Spécification

Spécification

MBL2
Polyclonal
Unconjugated
MBL2
COLEC1, Collectin 1, HSMBPC, Man binding protein, Mannose binding lectin, Mannose binding protein C, MBL, MBL2, MBP, MBP C, MBP1
Rabbit
Antigen Affinity Chromatography
RUO
4153
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11226
MBL2
MBL2 Fusion Protein Ag21508
150 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions