missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio MCM7 / CDC47 Mouse, Clone: SPM379, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 4176-MSM5X-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
MCM7 / CDC47 | |
Nuclear | |
SPM379 | |
Unconjugated | |
NP_005907 | |
Mouse | |
88kDa | |
Protein A/G Purified | |
RUO | |
4176 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
MCM7 | |
Recombinant human full-length hCDC47 protein | |
Exponentially growing any cultured human cells. Tonsil or lymph node. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |