missing translation for 'onlineSavingsMsg'
Learn More

MEST Mouse anti-Human, Mouse, Clone: 1A4A1, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 60263-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837315

  • 399.00€ / 150µL
Expédition estimée: 28-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the alpha/beta hydrolase superfamily. It is imprinted, exhibiting preferential expression from the paternal allele in fetal tissues, and isoform-specific imprinting in lymphocytes. The loss of imprinting of this gene has been linked to certain types of cancer and may be due to promotor switching. The encoded protein may play a role in development. Alternatively spliced transcript variants encoding multiple isoforms have been identified for this gene. Pseudogenes of this gene are located on the short arm of chromosomes 3 and 4, and the long arm of chromosomes 6 and 15.
Spécification

Spécification

MEST
Monoclonal
2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q07646, Q5EB52
Mest
MEST Fusion Protein Ag11929
150 μL
Primary
Human, Mouse
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1A4A1
Unconjugated
MEST
MEST, PEG1
Mouse
Protein A
RUO
17294, 4232
-20°C
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions