missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEST Mouse anti-Human, Mouse, Clone: 1A4A1, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | MEST |
---|---|
Clone | 1A4A1 |
Concentration | 2 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16847315
|
Proteintech
60263-1-IG-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 28-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16837315
|
Proteintech
60263-1-IG-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 28-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene encodes a member of the alpha/beta hydrolase superfamily. It is imprinted, exhibiting preferential expression from the paternal allele in fetal tissues, and isoform-specific imprinting in lymphocytes. The loss of imprinting of this gene has been linked to certain types of cancer and may be due to promotor switching. The encoded protein may play a role in development. Alternatively spliced transcript variants encoding multiple isoforms have been identified for this gene. Pseudogenes of this gene are located on the short arm of chromosomes 3 and 4, and the long arm of chromosomes 6 and 15.Spécification
MEST | |
2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MEST, PEG1 | |
Mest | |
IgG2a | |
Protein A | |
Antibody |
1A4A1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q07646, Q5EB52 | |
17294, 4232 | |
MEST Fusion Protein Ag11929 | |
Primary | |
-20°C | |
MEST |