missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEST Mouse anti-Human, Mouse, Clone: 1A4A1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60263-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the alpha/beta hydrolase superfamily. It is imprinted, exhibiting preferential expression from the paternal allele in fetal tissues, and isoform-specific imprinting in lymphocytes. The loss of imprinting of this gene has been linked to certain types of cancer and may be due to promotor switching. The encoded protein may play a role in development. Alternatively spliced transcript variants encoding multiple isoforms have been identified for this gene. Pseudogenes of this gene are located on the short arm of chromosomes 3 and 4, and the long arm of chromosomes 6 and 15.Spécification
MEST | |
Monoclonal | |
2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q07646, Q5EB52 | |
Mest | |
MEST Fusion Protein Ag11929 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2a |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
1A4A1 | |
Unconjugated | |
MEST | |
MEST, PEG1 | |
Mouse | |
Protein A | |
RUO | |
17294, 4232 | |
-20°C | |
Liquid |