missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10428-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a cell-surface glycoprotein found on melanoma cells. The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. The importance of the iron binding function has not yet been identified. This gene resides in the same region of chromosome 3 as members of the transferrin superfamily. Alternative splicing results in two transcript variants.Spécification
MFI2 | |
Polyclonal | |
Unconjugated | |
MELTF | |
CD228, MAP97, Melanotransferrin, MFI2, MTF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288038, 30060, 4241 | |
-20°C | |
Liquid |
Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08582, Q9R0R1 | |
Meltf | |
MFI2 Fusion Protein Ag0700 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |