missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICALL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24408-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
MILK2 may be a cytoskeletal regulator.Spécification
MICALL2 | |
Polyclonal | |
Unconjugated | |
MICALL2 | |
JRAB, MICAL L2, MICAL like 2, MICAL like protein 2, MICALL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79778 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IY33 | |
MICALL2 | |
MICALL2 Fusion Protein Ag19931 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |