Learn More
MMADHC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23191-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Involved in cobalamin metabolism.Spécification
MMADHC | |
Polyclonal | |
Unconjugated | |
MMADHC | |
C2orf25, cblD, CL25022, MMADHC, My011 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109129, 27249 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99LS1, Q9H3L0 | |
Mmadhc | |
MMADHC Fusion Protein Ag19343 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |