Learn More
CD1c Mouse anti-Human, PerCP-Cy5.5, Clone: F10/21A3, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 565424
Informations supplémentaires : Poids : 0.01000kg
Description
The F10/21A3 monoclonal antibody specifically binds to CD1c. The CD1 family of transmembrane glycoproteins are structurally related to the classical major histocompatibility complex (MHC) proteins. CD1c is a type I transmembrane glycoprotein that forms heterodimers with beta-2-microglobulin. CD1c presents lipids and glycolipids of self or microbial origin to T cells. CD1c is expressed by Langerhans cells, dendritic cells, monocytes, cortical thymocytes, T cells, and some B cells.
Spécification
CD1c | |
Monoclonal | |
PerCP-Cyanine5.5 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
GM-CSF- and IL-4-activated Human Monocytes | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
F10/21A3 | |
CD1; R7; M241; BDCA1 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
For Research Use Only.