missing translation for 'onlineSavingsMsg'
Learn More

NCOA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 20175-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829684

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Several alternatively spliced transcript variants have been found for this gene.
Spécification

Spécification

NCOA5
Polyclonal
Unconjugated
NCOA5
CIA, KIAA1637, NCoA 5, NCOA5, nuclear receptor coactivator 5
Rabbit
Antigen Affinity Chromatography
RUO
228869, 296372, 57727
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Flow Cytometry
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91W39, Q9HCD5
NCOA5
NCOA5 Fusion Protein Ag14014
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions