Learn More
NEDD4L Mouse anti-Human, Unconjugated, Clone: 2F6H6, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67276-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16829815
  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 
Description
Description
The NEDD4L antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NEDD4L. This antibody recognizes human, mouse, rat antigen. The NEDD4L antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
NEDD4L
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
ELISA, Western Blot
2F6H6
Unconjugated
20 μL
23327
IgG1