missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-H Mouse anti-Human, Porcine, Clone: 1A3C7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60331-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody recognize NEFH only.
Involved in the maintece of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).Spécification
NF-H | |
Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12036, P12037 | |
Mouse | |
Protein A | |
RUO | |
100156492, 4744 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
1A3C7 | |
Unconjugated | |
NEFH | |
NEFH | |
NF200 Fusion Protein Ag13517 | |
20 μL | |
Primary | |
Porcine, Human | |
Antibody | |
IgG2a |