Learn More
NFS1 Mouse anti-Human, Unconjugated, Clone: 3A10A7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67021-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
NFS1(nitrogen fixation 1 homolog) is also named as NIFS, HUSSY-08 and belongs to the class-V pyridoxal-phosphate-dependent aminotransferase family. The protein has been identified as a pyridoxal phosphate-containing homodimer that catalyzes the formation of equimolar amounts of elemental sulfur and L-alanine from the substrate, L-cysteine. It is reported that NFS1 is also able to catalyze the removal of selenium from selenocysteine, a mechanism similar to the L-cysteine reaction was postulated(PMID:9812986). It has 2 isoforms produced by alternative initiation.
Spécification
NFS1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, ELISA | |
3A10A7 | |
Unconjugated | |
20 μL | |
9054 | |
Liquid |