missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio NGF-Receptor (p75) / CD271 (Soft Tissue Tumor Marker) Mouse, Clone: NGFR/1964, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 4804-MSM4-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
NGF-Receptor (p75) / CD271 (Soft Tissue Tumor Marker) | |
Cell surface and cytoplasmic | |
NGFR/1964 | |
Unconjugated | |
Mouse | |
75kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
4804 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human p75 NGFR protein (apprx. aa 281-421) | |
Neuronal axons, Schwann cells, and perineural cells of peripheral nerves, or tumors of nerve sheath differentiation e.g. Schwannoma, Neurofibroma. Soma and axons of sensory neurons, and ganglionic satellite cells. Melano | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |