missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14380-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Plays a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. It is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.Spécification
NIT1 | |
Polyclonal | |
Unconjugated | |
NIT1 | |
NIT1, nitrilase 1, Nitrilase homolog 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27045, 289222, 4817 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TQ94, Q86X76, Q8VDK1 | |
Nit1 | |
NIT1 Fusion Protein Ag5764 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |