missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKRF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14693-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predomitly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.Spécification
NKRF | |
Polyclonal | |
Unconjugated | |
NKRF | |
ITBA4, NF kappa B repressing factor, NKRF, NRF, Protein ITBA4, Transcription factor NRF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100910054, 55922, 77286 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15226, Q8BY02 | |
NKRF | |
NKRF Fusion Protein Ag6252 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |