missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17215-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
NLR family member X1 (NLRX1) belongs to the NLRP family and contains four LRR (leucine-rich) repeats and one NATCH domain. NLRX1 localizes to the mitochondrial outer membrane and interacts through the mitochondrial antiviral signalling adaptor MAVS during viral infection. Expression of NLRX1 results in the potent inhibition of RLH- and MAVS-mediated interferon-beta promoter activity and in the disruption of virus-induced RLH-MAVS interactions. It is also reported that NLRX1 synergistically potentiated ROS production induced by tumour necrosis factor alpha, Shigella infection and double-stranded RNA, resulting in amplified NF-kappaB-dependent and JUN amino-terminal kinases-dependent signalling, thus contributing to the link between ROS generation at the mitochondria and innate immune responses.Spécification
NLRX1 | |
Polyclonal | |
Unconjugated | |
NLRX1 | |
Caterpiller protein 11.3, CLR11.3, DLNB26, NLR family member X1, NLRX1, NOD26, NOD5, NOD9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
270151, 79671 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TL44, Q86UT6 | |
NLRX1 | |
NLRX1 Fusion Protein Ag11000 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |