Learn More
NME3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15136-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
NME3 mRNA is preferentially expressed at early stages of myeloid differentiation of highly purified CD34(+) cells. Its constitutive expression in a myeloid precursor line, which is growth-factor dependent for both proliferation and differentiation, results in inhibition of granulocytic differentiation induced by granulocyte colony-stimulating factor and causes apoptotic cell death. These results appear consistent with a role for the NME3 gene in normal hematopoiesis and raise the possibility that its overexpression contributes to differentiation arrest, a feature of blastic transformation in chronic myelogenous leukemia.Spécification
NME3 | |
Polyclonal | |
Unconjugated | |
NME3 | |
c371H6.2, DR nm23, KIAA0516, NDK 3, NDP kinase 3, NDPK C, NDPKC, NME3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4832, 79059, 85269 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13232, Q9WV85 | |
NME3 | |
NME3 Fusion Protein Ag7284 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |