missing translation for 'onlineSavingsMsg'
Learn More

NRF1 Mouse anti-Human, Clone: 3C2B8, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66832-1-IG-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829075

  • 140.00€ / 20µL
Expédition estimée: 29-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for ""nuclear factor (erythroid-derived 2)-like 1"" which has an official symbol of NFE2L1.
Spécification

Spécification

NRF1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q16656
NRF1
NRF1 Fusion Protein Ag28149
20 μL
Primary
Human
Antibody
IgG2b
Western Blot
3C2B8
Unconjugated
NRF1
ALPHA PAL, NRF 1, NRF1, nuclear respiratory factor 1
Mouse
Protein A
RUO
4899
-20°C
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions