missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRF1 Mouse anti-Human, Clone: 3C2B8, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | NRF1 |
---|---|
Clone | 3C2B8 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16829075
|
Proteintech
66832-1-IG-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16819075
|
Proteintech
66832-1-IG-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for ""nuclear factor (erythroid-derived 2)-like 1"" which has an official symbol of NFE2L1.Spécification
NRF1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ALPHA PAL, NRF 1, NRF1, nuclear respiratory factor 1 | |
NRF1 | |
IgG2b | |
Protein A | |
Antibody |
3C2B8 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q16656 | |
4899 | |
NRF1 Fusion Protein Ag28149 | |
Primary | |
-20°C | |
NRF1 |