missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRF1 Mouse anti-Human, Clone: 3C2B8, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66832-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for ""nuclear factor (erythroid-derived 2)-like 1"" which has an official symbol of NFE2L1.Spécification
NRF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16656 | |
NRF1 | |
NRF1 Fusion Protein Ag28149 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
3C2B8 | |
Unconjugated | |
NRF1 | |
ALPHA PAL, NRF 1, NRF1, nuclear respiratory factor 1 | |
Mouse | |
Protein A | |
RUO | |
4899 | |
-20°C | |
Liquid |