missing translation for 'onlineSavingsMsg'
Learn More

ODAM Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16509-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836164

  • 140.00€ / 20µL
Expédition estimée: 29-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Tooth-associated epithelia protein that probably plays a role in odontogenesis, the complex process that results in the initiation and generation of the tooth. May be incorporated in the enamel matrix at the end of mineralization process. Involved in the induction of RHOA activity via interaction with ARHGEF and expression of downstream factors such as ROCK. Plays a role in attachment of the junctional epithelium to the tooth surface.
Spécification

Spécification

ODAM
Polyclonal
Unconjugated
ODAM
APIN, ODAM
Rabbit
Antigen Affinity Chromatography
RUO
54959
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A1E959
ODAM
ODAM Fusion Protein Ag9726
20 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions