missing translation for 'onlineSavingsMsg'
Learn More

ODF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24736-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16833225

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The outer dense fibers are cytoskeletal structures that surround the axoneme in the middle piece and principal piece of the sperm tail. The fibers function in maintaining the elastic structure and recoil of the sperm tail as well as in protecting the tail from shear forces during epididymal transport and ejaculation. Defects in the outer dense fibers lead to abnormal sperm morphology and infertility. The human outer dense fibers contains at least 10 major proteins and this gene encodes the main protein.
Spécification

Spécification

ODF1
Polyclonal
Unconjugated
ODF1
HSPB10, ODF, ODF1, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, Outer dense fiber protein 1, RT7, SODF
Rabbit
Antigen Affinity Chromatography
RUO
18285, 24610, 4956
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P21769, Q14990, Q61999
Odf1
ODF1 Fusion Protein Ag20562
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions