missing translation for 'onlineSavingsMsg'
Learn More

Orexin receptor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 18370-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16828444

  • 325.38€ / 150µL
Expédition estimée: 04-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.
Spécification

Spécification

Orexin receptor 1
Polyclonal
Unconjugated
HCRTR1
HCRTR1, hypocretin (orexin) receptor 1, Hypocretin receptor type 1, Orexin receptor 1, Orexin receptor type 1, Ox 1 R, Ox1 R, OX1R
Rabbit
Antigen Affinity Chromatography
RUO
230777, 25593, 3061
-20°C
Liquid
Western Blot, Flow Cytometry
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43613, P56718, P58307
Hcrtr1
Orexin receptor 1 Fusion Protein Ag12269
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions