missing translation for 'onlineSavingsMsg'
Learn More

P16-INK4A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10883-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837563

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

10883-1-AP reacts with p16 specifically. P16 plays an essential role in regulating the cell cycle, and mutations in p16 increase the risk of developing various cancers, including melanoma.

Kaiso is a novel member of the BTB/POZ (Broad complex, Tramtrak, Bric a brac/Pox virus and zinc finger) zinc finger superfamily of transcription factors. Yeast 2-hybrid studies have shown that Kaiso interacts with catenin p120ctn. This interaction involves the Arm repeats of 1-7 of the p120ctn Armadillo domain and the carboxyl-terminal third of Kaiso. The significance of the Kaiso:p120 ctn interaction is still unknown.
Spécification

Spécification

P16-INK4A
Polyclonal
Unconjugated
CDKN2A
ARF, ARF,CDKN2A, CDK4 inhibitor p16 INK4, CDK4 inhibitor p16INK4, CDK4I, CDKN2, CDKN2A, CMM2, INK4, INK4a, MLM, MTS 1, MTS1, Multiple tumor suppressor 1, P16, p16 INK4, p16 INK4a, p16INK4, p16INK4a, TP16
Rabbit
Antigen Affinity Chromatography
RUO
1029
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N726
CDKN2A
P16-INK4A Fusion Protein Ag1328
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions